Merck
  • Constitutive STAT3-activation in Sezary syndrome: tyrphostin AG490 inhibits STAT3-activation, interleukin-2 receptor expression and growth of leukemic Sezary cells.
Constitutive STAT3-activation in Sezary syndrome: tyrphostin AG490 inhibits STAT3-activation, interleukin-2 receptor expression and growth of leukemic Sezary cells.
Leukemia (2001-05-23)
K W Eriksen, K Kaltoft, G Mikkelsen, M Nielsen, Q Zhang, C Geisler, M H Nissen, C Röpke, M A Wasik, N Odum
ABSTRACT
Interleukin-2 (IL-2) is a growth factor which upon binding to high-affinity receptors (IL-2Ralphabetagamma) triggers mitogenesis in T cells. IL-2Ralpha expression is restricted to T cells which have recently encountered antigen, and in healthy individuals the majority (>95%) of peripheral T cells are IL-2Ralpha negative. An aberrant expression of IL-2Ralpha has recently been described in cutaneous T-cell lymphoma (CTCL). Here, we study the regulation of IL-2Ralpha expression and STATs in a tumor cell line obtained from peripheral blood from a patient with Sezary syndrome (SS), a leukemic variant of CTCL. We show that (1) STAT3 (a transcription factor known to regulate IL-2Ralpha transcription) is constitutively tyrosine-phosphorylated in SS tumor cells, but not in non-malignant T cells; (2) STAT3 binds constitutively to a STAT-binding sequence in the promotor of the IL-2Ralpha gene; (3) the Janus kinase inhibitor, tyrphostine AG490, inhibits STAT3 activation, STAT3 DNA binding, and IL-2Ralpha mRNA and protein expression in parallel; and (4) tyrphostine AG490 inhibits IL-2 driven mitogenesis and triggers apoptosis in SS tumor cells. In conclusion, we provide the first example of a constitutive STAT3 activation in SS tumor cells. Moreover, our findings suggest that STAT3 activation might play an important role in the constitutive IL-2Ralpha expression, survival, and growth of malignant SS cells.