Merck
Reporter Immobilization Assay (REIA) for Bioconjugating Reactions.
Bioconjugate chemistry (2016-05-18)
Martin Schatte, Marco Bocola, Teresa Roth, Ronny Martinez, Erhard Kopetzki, Ulrich Schwaneberg, Mara Bönitz-Dulat
ABSTRACT
Enzymes able to ligate biomolecules are emerging tools to generate site-specific bioconjugates. In this study we present a detection and screening method for bioconjugating enzymes which overcomes limitations of analytical methods such as HPLC or MS. These techniques are experimentally demanding and often limited in sensitivity and throughput compared to enzymatic assays. The principle of this Reporter Immobilization Assay (REIA) is the ligation of a reporter enzyme to a peptide carrying an affinity handle, which can be utilized for its isolation. The REIA system exhibits a high sensitivity with a linear range down to 1 μg/mL (55 nM), a variation coefficient of 6.5%, and can be performed cost-efficiently in 96-well microtiter plate format. The application of this assay allowed the characterization of a thiol transpeptidase sortase from S. aureus which is an important drug target and a biotechnological tool for ligation and modification of proteins. Thereby, yet-undetectable promiscuous activity of sortase could be detected, e.g., the acceptance of alanine as nucleophile. In addition, we were able to provide evidence that the REIA is suitable for high throughput screening of enzyme libraries using crude cellular extract with a throughput of 600 samples per hour.
Product Description
Roche
Streptavidin Magnetic Particles, suspension
Pricing and availability is not currently available.