Merck
  • Heme Oxygenase-1 Modulates Human Respiratory Syncytial Virus Replication and Lung Pathogenesis during Infection.
Heme Oxygenase-1 Modulates Human Respiratory Syncytial Virus Replication and Lung Pathogenesis during Infection.
Journal of immunology (Baltimore, Md. : 1950) (2017-06-02)
Janyra A Espinoza, Miguel A León, Pablo F Céspedes, Roberto S Gómez, Gisela Canedo-Marroquín, Sebastían A Riquelme, Francisco J Salazar-Echegarai, Phillipe Blancou, Thomas Simon, Ignacio Anegon, Margarita K Lay, Pablo A González, Claudia A Riedel, Susan M Bueno, Alexis M Kalergis
ABSTRACT
Human respiratory syncytial virus (hRSV) is the leading cause of severe lower respiratory tract infections in children. The development of novel prophylactic and therapeutic antiviral drugs against hRSV is imperative to control the burden of disease in the susceptible population. In this study, we examined the effects of inducing the activity of the host enzyme heme oxygenase-1 (HO-1) on hRSV replication and pathogenesis on lung inflammation induced by this virus. Our results show that after hRSV infection, HO-1 induction with metalloporphyrin cobalt protoporphyrin IX significantly reduces the loss of body weight due to hRSV-induced disease. Further, HO-1 induction also decreased viral replication and lung inflammation, as evidenced by a reduced neutrophil infiltration into the airways, with diminished cytokine and chemokine production and reduced T cell function. Concomitantly, upon cobalt protoporphyrin IX treatment, there is a significant upregulation in the production of IFN-α/β mRNAs in the lungs. Furthermore, similar antiviral and protective effects occur by inducing the expression of human HO-1 in MHC class II
Sigma-Aldrich
Bovine IFNG / Interferon Gamma ELISA Kit, for serum, plasma and cell culture supernatants
Pricing and availability is not currently available.