Merck
  • Profile of differentially expressed Toll-like receptor signaling genes in the natural killer cells of patients with Sézary syndrome.
Profile of differentially expressed Toll-like receptor signaling genes in the natural killer cells of patients with Sézary syndrome.
Oncotarget (2017-12-02)
Kelly C G ManfrereC, Marina P Torrealba, Denis R Miyashiro, Nátalli Z Pereira, Fabio S Y Yoshikawa, Luana de M Oliveira, Jade Cury-Martins, Alberto J S Duarte, José A Sanches, Maria N Sato
ABSTRACT
Sézary syndrome (SS), an aggressive and leukemic form of cutaneous T-cell lymphoma, usually results in shortened survival. Improving innate immunity in SS by targeting natural killer (NK) cells with Toll-like receptor (TLR) agonists could be an interesting modulatory strategy. We evaluated the NK cell populations in SS patients assessing activating and inhibitory receptors expression and profiled the differential expression of TLR signaling pathway genes in unstimulated NK cells and after TLR7/8 stimulation. We observed preserved CD56
Sigma-Aldrich
Human MICB ELISA Kit, for serum, plasma, cell culture supernatant and urine
Pricing and availability is not currently available.