Merck
LC/MS Analysis of Fructose and NaCl on SeQuant® ZIC-HILIC
LC/MS Analysis of Fructose and NaCl on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
SeQuant ZIC-cHILIC, PEEK, 150 x 4.6 mm, 5 μm (1.50461.0001)
mobile phase
[A] acetonitrile; [B] 100 mM Ammonium acetate, adjusted with acetic acid to pH 5.0; (80:20, A:B)
flow rate
1.5 mL/min
pressure
2000 psi 135 Bar
column temp.
40 °C
detector
ELSD (Analytical nebulizer)
injection
20 μL
sample
1000 ppm Fructose and 90 ppm NaCl (Diluent Acetonitrile and water 80:20 (v/v))
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany