Skip to Content
Merck

AABH14DCD3EC

AA BLOCKS, INC.

Strontium bromide

Synonym(s):

strontium dibromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/2BrH.Sr/h2*1H;/q;;+2/p-2

Inchi Key

YJPVTCSBVRMESK-UHFFFAOYSA-L

Purity

99%

Storage temp.

RT

Country of Origin

US



Still not finding the right product?

Explore all of our products under Strontium bromide




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library