Skip to Content
Merck

AABH97D2ABE4

AA BLOCKS, INC.

Nα-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride

Synonym(s):

(S)-N-(5-((diaminomethylene)amino)-1-((4-methyl-2-oxo-2H-chromen-7-yl)amino)-1-oxopentan-2-yl)benzamide hydrochloride

Select a Size

Change View

Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C23H25N5O4.ClH/c1-14-12-20(29)32-19-13-16(9-10-17(14)19)27-22(31)18(8-5-11-26-23(24)25)28-21(30)15-6-3-2-4-7-15;/h2-4,6-7,9-10,12-13,18H,5,8,11H2,1H3,(H,27,31)(H,28,30)(H4,24,25,26);1H/t18-;/m0./s1

Inchi Key

XKAVYOJOHZLTDF-FERBBOLQSA-N

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library