Skip to Content
Merck

AABH97D3CBBC

AA BLOCKS, INC.

Ethidium monoazide (bromide)

Synonym(s):

3-amino-8-azido-5-ethyl-6-phenylphenanthridinium bromide

Select a Size

Change View

About This Item

Linear Formula:
C21H18N5Br1
CAS Number:
Molecular Weight:
420.31
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C21H17N5.BrH/c1-2-26-20-12-15(22)8-10-18(20)17-11-9-16(24-25-23)13-19(17)21(26)14-6-4-3-5-7-14;/h3-13,22H,2H2,1H3;1H

Inchi Key

GHUXAYLZEGLXDA-UHFFFAOYSA-N

Purity

95%

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library