Sign In to View Organizational & Contract Pricing
Select a Size
Product AMBH996F2BA0 is not currently sold in your country.
About This Item
Product AMBH996F2BA0 is not currently sold in your country.
Iupac Name
6,7-dihydro-5H-pyrrolo[1,2-a]imidazol-6-ol
Inchi Code
1S/C6H8N2O/c9-5-3-6-7-1-2-8(6)4-5/h1-2,5,9H,3-4H2
InChI key
OLXNUSXUUWAEGE-UHFFFAOYSA-N
storage temp.
Sealed in dry, 2-8C
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/1378606-06-4.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/1378606-06-4.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
