Skip to Content
Merck

BDPH9BCC2FAF

Bide Pharmatech

5,5'-((2-Aminoethyl)azanediyl)bis(pentan-1-ol)

Synonym(s):

5,5'-((2-aminoethyl)azanediyl)bis(pentan-1-ol)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H28O2N2
CAS Number:
Molecular Weight:
232.37
MDL number:
Product BDPH9BCC2FAF is not currently sold in your country.


Inchi Code

1S/C12H28N2O2/c13-7-10-14(8-3-1-5-11-15)9-4-2-6-12-16/h15-16H,1-13H2

Inchi Key

NNZFGPKHQWRZSW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

90%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Colorless to Yellow Viscous Liquid or Semi-solid or lump



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library