Skip to Content
Merck

BDPH9BCC2FC7

Bide Pharmatech

Diethyl (aminomethyl)phosphonate

Synonym(s):

diethyl aminomethylphosphonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H14O3N1P1
CAS Number:
Molecular Weight:
167.14
MDL number:
Product BDPH9BCC2FC7 is not currently sold in your country.


Inchi Code

1S/C5H14NO3P/c1-3-8-10(7,5-6)9-4-2/h3-6H2,1-2H3

Inchi Key

UIBCDEFKKLRXHR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under Diethyl (aminomethyl)phosphonate



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library