Skip to Content
Merck

BDPH9BCC53FA

Bide Pharmatech

1-(4-Iodophenyl)propan-1-one

Synonym(s):

1-(4-iodophenyl)-1-propanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H9O1I1
CAS Number:
Molecular Weight:
260.07
MDL number:
Product BDPH9BCC53FA is not currently sold in your country.


Inchi Code

1S/C9H9IO/c1-2-9(11)7-3-5-8(10)6-4-7/h3-6H,2H2,1H3

Inchi Key

FJTCRHXSVHRKBR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 1-(4-Iodophenyl)propan-1-one


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library