Skip to Content
Merck

BDPH9BCC72BC

Bide Pharmatech

1-(3-Amino-5-fluoro-2-methylphenyl)ethanone

Synonym(s):

1-(3-amino-5-fluoro-2-methylphenyl)ethan-1-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H10O1N1F1
CAS Number:
Molecular Weight:
167.18
MDL number:
Product BDPH9BCC72BC is not currently sold in your country.


Inchi Code

1S/C9H10FNO/c1-5-8(6(2)12)3-7(10)4-9(5)11/h3-4H,11H2,1-2H3

Inchi Key

HLPQMTWLINPPLV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library