Skip to Content
Merck

BDPH9BCC90FA

Bide Pharmatech

2-Amino-2-[2-(trifluoromethyl)phenyl]ethan-1-ol

Synonym(s):

2-amino-2-[2-(trifluoromethyl)phenyl]ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H10O1N1F3
CAS Number:
Molecular Weight:
205.18
MDL number:
Product BDPH9BCC90FA is not currently sold in your country.


Inchi Code

1S/C9H10F3NO/c10-9(11,12)7-4-2-1-3-6(7)8(13)5-14/h1-4,8,14H,5,13H2

Inchi Key

OIRSHTSIVLNGMS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Keep in dark place, inert atmosphere, 2-8°C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library