Product BDPH9BCCBCD4 is not currently sold in your country.
Inchi Code
1S/C18H14OS.ClH/c19-15-11-13-18(14-12-15)20(16-7-3-1-4-8-16)17-9-5-2-6-10-17;/h1-14H;1H
Inchi Key
YAFVTOLUUZOHAS-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (4-Hydroxyphenyl)diphenylsulfonium chloride
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
