Skip to Content
Merck

BDPH9BCCD321

Bide Pharmatech

1-(4-Bromo-2-fluorophenyl)ethanamine

Synonym(s):

1-(4-bromo-2-fluorophenyl)ethanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9N1Br1F1
CAS Number:
Molecular Weight:
218.07
MDL number:
Product BDPH9BCCD321 is not currently sold in your country.


Inchi Code

1S/C8H9BrFN/c1-5(11)7-3-2-6(9)4-8(7)10/h2-5H,11H2,1H3

Inchi Key

BRPFKIPSXUHGSH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Liquid



Still not finding the right product?


hstatements

signalword

Danger

pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library