Skip to Content
Merck

BDPH9BCCDCE2

Bide Pharmatech

(4-(Phenanthren-9-yl)phenyl)boronic acid

Synonym(s):

(4-(phenanthren-9-yl)phenyl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H15O2B1
CAS Number:
Molecular Weight:
298.15
MDL number:
Product BDPH9BCCDCE2 is not currently sold in your country.


Inchi Code

1S/C20H15BO2/c22-21(23)16-11-9-14(10-12-16)20-13-15-5-1-2-6-17(15)18-7-3-4-8-19(18)20/h1-13,22-23H

Inchi Key

BRMXCUCHGKWTIO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library