Skip to Content
Merck

BDPH9BCCFA50

Bide Pharmatech

Methyl 2,3-difluoro-6-methylbenzoate

Synonym(s):

methyl 2,3-difluoro-6-methylbenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H8O2F2
CAS Number:
Molecular Weight:
186.16
MDL number:
Product BDPH9BCCFA50 is not currently sold in your country.


Inchi Code

1S/C9H8F2O2/c1-5-3-4-6(10)8(11)7(5)9(12)13-2/h3-4H,1-2H3

Inchi Key

BLZRUSJWDBBFIX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library