Skip to Content
Merck

BDPH9BCCFCC2

Bide Pharmatech

6-(Methoxycarbonyl)-3-methyl-1H-indole-2-carboxylic acid

Synonym(s):

6-(methoxycarbonyl)-3-methyl-1H-indole-2-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H11O4N1
CAS Number:
Molecular Weight:
233.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H11NO4/c1-6-8-4-3-7(12(16)17-2)5-9(8)13-10(6)11(14)15/h3-5,13H,1-2H3,(H,14,15)

Inchi Key

AYNZBBDTXXBQBM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library