Skip to Content
Merck

BDPH9BCCFF8B

Bide Pharmatech

3-Bromo-2-chloro-N-(1H-pyrazol-4-yl)benzamide

Synonym(s):

3-bromo-2-chloro-N-(1H-pyrazol-4-yl)benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H7O1N3Cl1Br1
CAS Number:
Molecular Weight:
300.54
MDL number:
Product BDPH9BCCFF8B is not currently sold in your country.


Inchi Code

1S/C10H7BrClN3O/c11-8-3-1-2-7(9(8)12)10(16)15-6-4-13-14-5-6/h1-5H,(H,13,14)(H,15,16)

Inchi Key

LTWGFZLXRDQRPW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library