Product BDPH9BCD014E is not currently sold in your country.
Inchi Code
1S/C23H32N2O2/c1-15(2)19-13-18(27-17-11-9-8-10-12-17)14-20(16(3)4)21(19)24-22(26)25-23(5,6)7/h8-16H,1-7H3,(H2,24,25,26)
Inchi Key
WLIVADBOJWNAPY-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 1-(tert-Butyl)-3-(2,6-diisopropyl-4-phenoxyphenyl)urea
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
