Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C6H9NO3S/c8-5-1-2-11-3-4(7-5)6(9)10/h4H,1-3H2,(H,7,8)(H,9,10)
Inchi Key
JICTZUFURQRLDW-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 5-Oxo-1,4-thiazepane-3-carboxylic acid
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
