Skip to Content
Merck

BDPH9BCD29F0

Bide Pharmatech

Ethyl 5-methylnicotinate

Synonym(s):

ethyl 5-methylnicotinate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O2N1
CAS Number:
Molecular Weight:
165.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11NO2/c1-3-12-9(11)8-4-7(2)5-10-6-8/h4-6H,3H2,1-2H3

Inchi Key

CLGKEUKISVVXPZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under Ethyl 5-methylnicotinate


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library