Skip to Content
Merck

BDPH9BCD2FD1

Bide Pharmatech

Ethyl 5-amino-4-bromo-3-methylthiophene-2-carboxylate

Synonym(s):

ethyl 5-amino-4-bromo-3-methylthiophene-2-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H10O2N1Br1S1
CAS Number:
Molecular Weight:
264.14
MDL number:
Product BDPH9BCD2FD1 is not currently sold in your country.


Inchi Code

1S/C8H10BrNO2S/c1-3-12-8(11)6-4(2)5(9)7(10)13-6/h3,10H2,1-2H3

Inchi Key

PMPBGORMHXRHRT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library