Product BDPH9BCD30E2 is not currently sold in your country.
Inchi Code
1S/C24H16N2/c1-3-11-19-17(7-1)23-15(9-5-13-21(23)25-19)16-10-6-14-22-24(16)18-8-2-4-12-20(18)26-22/h1-14,25-26H
Inchi Key
GDEFUYVXDBBUEJ-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Explore all of our products under 9H,9'H-4,4'-Bicarbazole
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
