Skip to Content
Merck

BDPH9BCD3ADE

Bide Pharmatech

11-Deoxymogroside V

Synonym(s):

(2S,3R,4S,5S,6R)-2-(((2S,3R,4S,5S,6R)-4,5-dihydroxy-2-(((3R,6R)-2-hydroxy-2-methyl-6-((3S,8R,9R,10S,13R,14S,17R)-4,4,9,13,14-pentamethyl-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)heptan-3-yl)oxy)-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C60H102O28
CAS Number:
Molecular Weight:
1271.45
MDL number:
Product BDPH9BCD3ADE is not currently sold in your country.


Inchi Code

1S/C60H102O28/c1-24(9-13-35(57(4,5)78)87-55-50(88-54-49(77)43(71)38(66)30(21-63)83-54)45(73)40(68)32(85-55)23-80-52-47(75)42(70)37(65)29(20-62)82-52)25-15-16-60(8)33-12-10-26-27(58(33,6)17-18-59(25,60)7)11-14-34(56(26,2)3)86-53-48(76)44(72)39(67)31(84-53)22-79-51-46(74)41(69)36(64)28(19-61)81-51/h10,24-25,27-55,61-78H,9,11-23H2,1-8H3/t24-,25-,27-,28-,29-,30-,31-,32-,33-,34+,35-,36-,37-,38-,39-,40-,41+,42+,43+,44+,45+,46-,47-,48-,49-,50-,51-,52-,53+,54+,55+,58+,59-,60+/m1/s1

Inchi Key

FXMONPOKYCDPKD-KUWQHOJOSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 11-Deoxymogroside V



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library