Skip to Content
Merck

BDPH9BCD3F06

Bide Pharmatech

2-Fluoro-3-isopropoxybenzoic acid

Synonym(s):

2-fluoro-3-isopropoxybenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H11O3F1
CAS Number:
Molecular Weight:
198.19
MDL number:
Product BDPH9BCD3F06 is not currently sold in your country.


Inchi Code

1S/C10H11FO3/c1-6(2)14-8-5-3-4-7(9(8)11)10(12)13/h3-6H,1-2H3,(H,12,13)

Inchi Key

ADISUGFAMDFUHD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?

Explore all of our products under 2-Fluoro-3-isopropoxybenzoic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library