Skip to Content
Merck

BDPH9BCD3FF3

Bide Pharmatech

Thiol-peg6-alcohol

Synonym(s):

17-mercapto-3,6,9,12,15-pentaoxaheptadecan-1-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H26O6S1
CAS Number:
Molecular Weight:
298.4
MDL number:
Product BDPH9BCD3FF3 is not currently sold in your country.


Inchi Code

1S/C12H26O6S/c13-1-2-14-3-4-15-5-6-16-7-8-17-9-10-18-11-12-19/h13,19H,1-12H2

Inchi Key

RBHVCADZZSNWRE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under Thiol-peg6-alcohol


hstatements

signalword

Danger

pictograms

CorrosionExclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library