Skip to Content
Merck

BDPH9BCD4E5F

Bide Pharmatech

(5-(((tert-Butoxycarbonyl)amino)methyl)thiophen-3-yl)boronic acid

Synonym(s):

(5-(((tert-butoxycarbonyl)amino)methyl)thiophen-3-yl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H16O4N1S1B1
CAS Number:
Molecular Weight:
257.12
MDL number:
Product BDPH9BCD4E5F is not currently sold in your country.


Inchi Code

1S/C10H16BNO4S/c1-10(2,3)16-9(13)12-5-8-4-7(6-17-8)11(14)15/h4,6,14-15H,5H2,1-3H3,(H,12,13)

Inchi Key

IGJFBQDBUNTHII-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library