Skip to Content
Merck

BDPH9BCD4F45

Bide Pharmatech

2,3,5,6-Tetrahydrobenzo[b][1,4,7]trioxonin-9-amine

Synonym(s):

2,3,5,6-tetrahydro-1,4,7-benzotrioxonin-9-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H13O3N1
CAS Number:
Molecular Weight:
195.22
MDL number:
Product BDPH9BCD4F45 is not currently sold in your country.


Inchi Code

1S/C10H13NO3/c11-8-1-2-9-10(7-8)14-6-4-12-3-5-13-9/h1-2,7H,3-6,11H2

Inchi Key

OOIADRPMMIAFGY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid or semi-solid or solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library