Skip to Content
Merck

BDPH9BCD693D

Bide Pharmatech

Methyl 4-amino-2-ethylbenzoate

Synonym(s):

methyl 4-amino-2-ethylbenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H13O2N1
CAS Number:
Molecular Weight:
179.22
MDL number:
Product BDPH9BCD693D is not currently sold in your country.


Inchi Code

1S/C10H13NO2/c1-3-7-6-8(11)4-5-9(7)10(12)13-2/h4-6H,3,11H2,1-2H3

Inchi Key

GUDJDBHCUBLRBQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

White to off-white solid

Storage temp.

Keep in dark place, inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under Methyl 4-amino-2-ethylbenzoate


hstatements

pcodes

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library