Skip to Content
Merck

BDPH9BCD71D9

Bide Pharmatech

4-Chloro-6-Fluoroindan-1-one

Synonym(s):

4-chloro-6-fluoro-1-indanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H6O1Cl1F1
Molecular Weight:
184.6
MDL number:
Product BDPH9BCD71D9 is not currently sold in your country.


Inchi Code

1S/C9H6ClFO/c10-8-4-5(11)3-7-6(8)1-2-9(7)12/h3-4H,1-2H2

Inchi Key

PADRGRWYNJBMQI-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library