Skip to Content
Merck

BDPH9BCD7D85

Bide Pharmatech

2-Amino-N-methylpropanamide hydrochloride

Synonym(s):

2-amino-N-methylpropanamide hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H11O1N2Cl1
CAS Number:
Molecular Weight:
138.6
MDL number:
Product BDPH9BCD7D85 is not currently sold in your country.


Inchi Code

1S/C4H10N2O.ClH/c1-3(5)4(7)6-2;/h3H,5H2,1-2H3,(H,6,7);1H

Inchi Key

UHWFDFLWTAIHRE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library