Skip to Content
Merck

BDPH9BCD94CD

Bide Pharmatech

N-((3-(4-Fluorophenyl)propanoyl)oxy)methanesulfonamide

Synonym(s):

N-((3-(4-fluorophenyl)propanoyl)oxy)methanesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12O4N1S1F1
CAS Number:
Molecular Weight:
261.27
MDL number:
Product BDPH9BCD94CD is not currently sold in your country.


Inchi Code

1S/C10H12FNO4S/c1-17(14,15)12-16-10(13)7-4-8-2-5-9(11)6-3-8/h2-3,5-6,12H,4,7H2,1H3

Inchi Key

UQXJGIBNZMIJEF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

signalword

Danger

pictograms

FlameCorrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library