Skip to Content
Merck

BDPH9BCDA080

Bide Pharmatech

Copper(II) isobutyrate

Synonym(s):

copper(2+) bis(2-methylpropanoate)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H14O4Cu1
CAS Number:
Molecular Weight:
237.74
MDL number:
Product BDPH9BCDA080 is not currently sold in your country.


Inchi Code

1S/2C4H8O2.Cu/c2*1-3(2)4(5)6;/h2*3H,1-2H3,(H,5,6);/q;;+2/p-2

Inchi Key

KOKFUFYHQQCNNJ-UHFFFAOYSA-L

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Blue powder



Still not finding the right product?

Explore all of our products under Copper(II) isobutyrate



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library