Skip to Content
Merck

BDPH9BCDAAE2

Bide Pharmatech

tert-Butyl 8-oxo-5-azaspiro[2.5]octane-5-carboxylate

Synonym(s):

tert-butyl 8-oxo-5-azaspiro[2.5]octane-5-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H19O3N1
CAS Number:
Molecular Weight:
225.29
MDL number:
Product BDPH9BCDAAE2 is not currently sold in your country.


Inchi Code

1S/C12H19NO3/c1-11(2,3)16-10(15)13-7-4-9(14)12(8-13)5-6-12/h4-8H2,1-3H3

Inchi Key

IEHDYRJCLKTFQY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

White to Yellow Solid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library