Skip to Content
Merck

BDPH9BCDABDC

Bide Pharmatech

1,8-Diazaspiro[4.5]decane dihydrochloride

Synonym(s):

1,8-diazaspiro[4.5]decane dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H18N2Cl2
CAS Number:
Molecular Weight:
213.15
MDL number:
Product BDPH9BCDABDC is not currently sold in your country.


Inchi Code

1S/C8H16N2.2ClH/c1-2-8(10-5-1)3-6-9-7-4-8;;/h9-10H,1-7H2;2*1H

Inchi Key

UPHOLJXKVDTBGG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to yellow powder or crystals



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library