Skip to Content
Merck

BDPH9BCDB280

Bide Pharmatech

(2-Methyl-1,3-dioxolan-2-yl)methanamine

Synonym(s):

(2-methyl-1,3-dioxolan-2-yl)methanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H11O2N1
Molecular Weight:
117.15
MDL number:
Product BDPH9BCDB280 is not currently sold in your country.


Inchi Code

1S/C5H11NO2/c1-5(4-6)7-2-3-8-5/h2-4,6H2,1H3

Inchi Key

BVVSUFVLGBJYOR-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library