Skip to Content
Merck

Skip To

BDPH9BCDB576

Bide Pharmatech

(4-Acrylamidophenyl)boronic acid

Synonym(s):

(4-acrylamidophenyl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H10O3N1B1
CAS Number:
Molecular Weight:
190.99
MDL number:
Product BDPH9BCDB576 is not currently sold in your country.


Inchi Code

1S/C9H10BNO3/c1-2-9(12)11-8-5-3-7(4-6-8)10(13)14/h2-6,13-14H,1H2,(H,11,12)

Inchi Key

JILCNOLYVKMITP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under (4-Acrylamidophenyl)boronic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library