Skip to Content
Merck

BDPH9BCDBB93

Bide Pharmatech

2-(2-Isopropoxyphenyl)acetaldehyde

Synonym(s):

2-(2-isopropoxyphenyl)acetaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H14O2
CAS Number:
Molecular Weight:
178.23
MDL number:
Product BDPH9BCDBB93 is not currently sold in your country.


Inchi Code

1S/C11H14O2/c1-9(2)13-11-6-4-3-5-10(11)7-8-12/h3-6,8-9H,7H2,1-2H3

Inchi Key

LCNAMKFQUIFNEJ-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 2-(2-Isopropoxyphenyl)acetaldehyde



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library