Skip to Content
Merck

BDPH9BCDBC32

Bide Pharmatech

4-Bromo-3-cyanopyridine

Synonym(s):

4-bromonicotinonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H3N2Br1
CAS Number:
Molecular Weight:
183.01
MDL number:
Product BDPH9BCDBC32 is not currently sold in your country.


Inchi Code

1S/C6H3BrN2/c7-6-1-2-9-4-5(6)3-8/h1-2,4H

Inchi Key

DMQBJRMKGVMOHX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Form: solid

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under 4-Bromo-3-cyanopyridine


signalword

Danger

pictograms

CorrosionSkull and crossbones


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library