Skip to Content
Merck

BDPH9BCDC1E1

Bide Pharmatech

Cyclopropanecarbaldehyde

Synonym(s):

cyclopropanecarbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H6O1
Molecular Weight:
70.09
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H6O/c5-3-4-1-2-4/h3-4H,1-2H2

Inchi Key

JMYVMOUINOAAPA-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library