Product BDPH9BCDCD30 is not currently sold in your country.
Inchi Code
1S/C13H16O3/c1-9(14)10-6-5-7-11(8-10)12(15)16-13(2,3)4/h5-8H,1-4H3
Inchi Key
YESSMNWGVXXGJB-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Viscous liquid
Still not finding the right product?
Explore all of our products under tert-Butyl 3-acetylbenzoate
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
