Product BDPH9BCDCD5D is not currently sold in your country.
Inchi Code
1S/C12H15ClO2S/c13-16(14,15)12-8-6-11(7-9-12)10-4-2-1-3-5-10/h6-10H,1-5H2
Inchi Key
CREMYEDHKUWVTB-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 4-Cyclohexylbenzene-1-sulfonyl chloride
hstatements
signalword
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
