Product BDPH9BCDCFCB is not currently sold in your country.
Inchi Code
1S/C6H3BrClN3/c7-3-1-9-4-2-10-6(8)11-5(3)4/h1-2,9H
Inchi Key
XVPYDBHFZKKIDV-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Country of Origin
CN
Physical Form
Solid
Storage temp.
Inert atmosphere, 2-8°C
Still not finding the right product?
Explore all of our products under 7-Bromo-2-chloro-5H-pyrrolo[3,2-d]pyrimidine
hstatements
signalword
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
