Skip to Content
Merck

BDPH9BCDDDF1

Bide Pharmatech

Methyl 2-acetyl-3-hydroxybenzoate

Synonym(s):

methyl 2-acetyl-3-hydroxybenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H10O4
CAS Number:
Molecular Weight:
194.19
MDL number:
Product BDPH9BCDDDF1 is not currently sold in your country.


Inchi Code

1S/C10H10O4/c1-6(11)9-7(10(13)14-2)4-3-5-8(9)12/h3-5,12H,1-2H3

Inchi Key

YLGAWYQBSGMTLX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under Methyl 2-acetyl-3-hydroxybenzoate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library