Skip to Content
Merck

BDPH9BCDE22B

Bide Pharmatech

2-(4-(Cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Synonym(s):

2-(4-(cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H23O3B1
CAS Number:
Molecular Weight:
274.17
MDL number:
Product BDPH9BCDE22B is not currently sold in your country.


Storage temp.

Inert atmosphere, 2-8°C

Inchi Code

1S/C16H23BO3/c1-15(2)16(3,4)20-17(19-15)13-7-9-14(10-8-13)18-11-12-5-6-12/h7-10,12H,5-6,11H2,1-4H3

Inchi Key

NEQVQJCPHVVIFO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library