Skip to Content
Merck

BDPH9BCDEFBB

Bide Pharmatech

6-Hydroxy-3-methylindazole

Synonym(s):

3-methyl-1H-indazol-6-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H8O1N2
CAS Number:
Molecular Weight:
148.16
MDL number:
Product BDPH9BCDEFBB is not currently sold in your country.


Inchi Code

1S/C8H8N2O/c1-5-7-3-2-6(11)4-8(7)10-9-5/h2-4,11H,1H3,(H,9,10)

Inchi Key

WTNPPQUZRFSUQF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Powder or crystals

Storage temp.

Keep in dark place, inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under 6-Hydroxy-3-methylindazole


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library