Skip to Content
Merck

BDPH9BCDF033

Bide Pharmatech

6-Cyano-1-tetralone

Synonym(s):

5-oxo-5,6,7,8-tetrahydro-2-naphthalenecarbonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H9O1N1
CAS Number:
Molecular Weight:
171.2
MDL number:
Product BDPH9BCDF033 is not currently sold in your country.


Inchi Code

1S/C11H9NO/c12-7-8-4-5-10-9(6-8)2-1-3-11(10)13/h4-6H,1-3H2

Inchi Key

FGWRKZJKRYCDOF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Yellow to brown to yellow-brown powder or crystals



Still not finding the right product?

Explore all of our products under 6-Cyano-1-tetralone


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library