Skip to Content
Merck

BDPH9BCDF22C

Bide Pharmatech

5-Fluoropyridine-2,3-dicarboxylic acid hydrochloride

Synonym(s):

5-fluoropyridine-2,3-dicarboxylic acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5O4N1Cl1F1
CAS Number:
Molecular Weight:
221.57
MDL number:
Product BDPH9BCDF22C is not currently sold in your country.


Inchi Code

1S/C7H4FNO4.ClH/c8-3-1-4(6(10)11)5(7(12)13)9-2-3;/h1-2H,(H,10,11)(H,12,13);1H

Inchi Key

OJTIFQVFXTXONN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library